Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zgpat | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001048148.1 | NP_001041613.1 | 3 | 994 | ||
NM_144894.3 | NP_659143.1 | 4 | 1218 | |||
GenBank mRNA | AK041491.1 | - | 4 | 1222 | ||
AK049764.1 | - | 3 | 993 | |||
AK052236.1 | - | 3 | 892 | |||
AK159801.1 | - | 3 | 1013 | |||
AK209697.1 | - | 2 | 71 | |||
BC021513.1 | - | 3 | 992 | 5351756 | ||
BC027218.1 | - | 3 | 901 | 3158198 |