Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ythdf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145919.1 | NP_001139391.1 | 4 | 1857 | ||
NM_172677.3 | NP_766265.3 | 5 | 1890 | |||
GenBank mRNA | AK039176.1 | - | 5 | 1953 | ||
AK053736.1 | - | 5 | 1925 | |||
AK078922.1 | - | 1 | 625 | |||
AK137245.1 | - | 5 | 1837 | |||
BC022932.1 | - | 1 | 315 | |||
BC042627.1 | - | 1 | 1407 | |||
BC052631.1 | - | 1 | 1207 | |||
BC057158.1 | - | 5 | 1917 | 5348717 | ||
BC067040.1 | - | 4 | 1828 | 6853196 | ||
BC067042.1 | - | 5 | 1823 | 6856248 |