Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acad9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172678.3 | NP_766266.3 | 11 | 1177 | ||
GenBank mRNA | AK028571.1 | - | 10 | 1891 | ||
AK031820.1 | - | 11 | 1109 | |||
AK075984.1 | - | 11 | 1096 | |||
AK145423.1 | - | 11 | 1085 | |||
AK212715.1 | - | 2 | 68 | |||
BC004737.1 | - | 5 | 423 | |||
BC031137.1 | - | 11 | 1113 | 4971848 | ||
BC032213.1 | - | 11 | 1118 | 5323723 | ||
BC033277.1 | - | 11 | 1098 | 4241041 |