Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hnrnpa3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053263.1 | NP_444493.1 | 4 | 459 | ||
NM_146130.3 | NP_666242.2 | 4 | 525 | |||
NM_198090.2 | NP_932758.1 | 4 | 525 | |||
GenBank mRNA | AB201711.1 | - | 4 | 505 | ||
AF463524.1 | - | 4 | 437 | |||
AK010481.1 | - | 4 | 453 | |||
AK133860.1 | - | 4 | 524 | |||
AK169663.1 | - | 4 | 507 | |||
BC023828.1 | - | 4 | 478 | 5343619 | ||
BC023908.1 | - | 4 | 494 | 5322033 | ||
BC024454.1 | - | 1 | 42 | |||
BC038364.1 | - | 4 | 507 | 5362605 | ||
BC057655.1 | - | 4 | 438 | |||
BC062198.1 | - | 4 | 516 | 2655879 | ||
BC064824.1 | - | 4 | 490 | 6477672 | ||
BC085474.1 | - | 3 | 400 | 30538435 | ||
BC092083.1 | - | 1 | 120 | |||
BC116250.1 | - | 4 | 428 | 40093600 | ||
BC116251.1 | - | 4 | 428 | 40093602 | ||
BC157919.1 | - | 4 | 428 | 9007347 | ||
BC158038.1 | - | 4 | 428 | 9007341 | ||
CO045618.1 | - | 3 | 309 |
Gene Symbol: Gm6793 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC157904.1 | - | 1 | 428 | 9088095 | |
BC158074.1 | - | 1 | 428 | 9007371 | ||
BC172011.1 | - | 1 | 428 | 9054297 |