Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpr149 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177346.4 | NP_796320.2 | 1 | 644 | ||
GenBank mRNA | AK136826.1 | - | 1 | 645 |
Set Membership: |
G-protein coupled receptor |