Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpr149 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177346.4 | NP_796320.2 | 3 | 1776 | ||
GenBank mRNA | AK035931.1 | - | 3 | 339 | ||
AK048373.1 | - | 499 | ||||
AK136826.1 | - | 3 | 1777 | |||
AK166029.1 | - | 604 | ||||
BC119599.2 | - | 3 | 440 |
Set Membership: |
G-protein coupled receptor |