Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fcrl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136236.1 | NP_001129708.1 | 9 | 955 | ||
NM_153090.2 | NP_694730.2 | 11 | 1144 | |||
NM_178165.4 | NP_835459.1 | 10 | 1084 | |||
GenBank mRNA | AF329485.1 | - | 10 | 1011 | ||
AK036445.1 | - | 5 | 1320 | |||
AK038494.1 | - | 10 | 1024 | |||
AK041750.1 | - | 11 | 1145 | |||
AK156528.1 | - | 10 | 1025 | |||
AK156985.1 | - | 9 | 895 | |||
AY158088.1 | - | 10 | 1004 | |||
AY506554.1 | - | 10 | 1017 | |||
AY506555.1 | - | 9 | 888 |