Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Isg20l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177663.4 | NP_808331.1 | 3 | 1260 | ||
GenBank mRNA | AK053778.1 | - | 3 | 1261 | ||
AK076747.1 | - | 3 | 983 | |||
AK155979.1 | - | 3 | 1264 |