Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc25a44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145876.2 | NP_001139348.1 | 2 | 652 | ||
NM_001145877.2 | NP_001139349.1 | 2 | 698 | |||
NM_001281795.1 | NP_001268724.1 | 2 | 573 | |||
NM_178696.5 | NP_848811.2 | 2 | 698 | |||
GenBank mRNA | AK032960.1 | - | 2 | 676 | ||
AK044648.1 | - | 2 | 545 | |||
AK046586.1 | - | 2 | 661 | |||
AK079532.1 | - | 2 | 662 | |||
AK083273.1 | - | 2 | 668 | |||
AK083470.1 | - | 2 | 548 | |||
AK134914.1 | - | 2 | 697 | |||
AK144731.1 | - | 2 | 624 | |||
AK165159.1 | - | 2 | 665 | |||
AK166011.1 | - | 2 | 670 | |||
AK166331.1 | - | 2 | 659 | |||
AK168955.1 | - | 2 | 671 | |||
AK169886.1 | - | 2 | 662 | |||
BC012234.1 | - | 2 | 477 | 4191504 | ||
BC052771.1 | - | 2 | 643 | 6310271 |