Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dennd4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_201407.4 | NP_958809.3 | 22 | 3712 | ||
GenBank mRNA | AK122294.1 | - | 9 | 2559 | ||
AK155638.1 | - | 22 | 3694 | |||
AY512564.1 | - | 21 | 3284 | |||
BC026996.1 | - | 3 | 562 | |||
BC040460.1 | - | 11 | 1759 | |||
BC049972.1 | - | 12 | 1998 | |||
BC066102.1 | - | 20 | 3146 | 30531856 | ||
BC151194.1 | - | 21 | 3358 | 9088082 |