Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dennd4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_201407.4 | NP_958809.3 | 19 | 3052 | ||
GenBank mRNA | AK122294.1 | - | 7 | 1410 | ||
AK155638.1 | - | 19 | 3034 | |||
AY512564.1 | - | 18 | 2624 | |||
BC040460.1 | - | 8 | 1099 | |||
BC049972.1 | - | 9 | 1338 | |||
BC066102.1 | - | 17 | 2486 | 30531856 | ||
BC151194.1 | - | 18 | 2698 | 9088082 |