Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Csde1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161854.1 | NP_001155326.1 | 12 | 1833 | ||
NM_144901.3 | NP_659150.1 | 13 | 1926 | |||
GenBank mRNA | AK122393.1 | - | 12 | 1743 | ||
AK144097.1 | - | 12 | 1772 | |||
AK145351.1 | - | 12 | 1938 | |||
AK145428.1 | - | 12 | 1755 | |||
AK145932.1 | - | 12 | 1750 | |||
AK146586.1 | - | 12 | 1774 | |||
AK146706.1 | - | 13 | 1858 | |||
AK147012.1 | - | 12 | 1772 | |||
AK150882.1 | - | 12 | 1751 | |||
AK151638.1 | - | 12 | 1769 | |||
AK151723.1 | - | 12 | 1773 | |||
AK151970.1 | - | 12 | 1773 | |||
AK152531.1 | - | 12 | 1769 | |||
AK152777.1 | - | 8 | 980 | |||
AK153196.1 | - | 12 | 1750 | |||
AK153263.1 | - | 8 | 980 | |||
AK154068.1 | - | 12 | 1758 | |||
AK155751.1 | - | 13 | 1851 | |||
AK159258.1 | - | 12 | 1766 | |||
AK159863.1 | - | 12 | 1766 | |||
AK168556.1 | - | 11 | 1569 | |||
BC016898.1 | - | 12 | 1770 | 4224466 | ||
BC024826.1 | - | 9 | 1053 | |||
BC029058.1 | - | 7 | 712 | 4947555 | ||
BC029688.1 | - | 12 | 1769 | 5367783 | ||
BC031404.1 | - | 12 | 1741 | 4953772 | ||
BC036720.1 | - | 9 | 1059 | |||
BC055072.1 | - | 2769 | ||||
BC062097.1 | - | 13 | 1841 | 6833505 |