Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Csde1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161854.1 | NP_001155326.1 | 18 | 2769 | ||
NM_144901.3 | NP_659150.1 | 19 | 2862 | |||
GenBank mRNA | AK122393.1 | - | 18 | 2679 | ||
AK144097.1 | - | 18 | 2709 | |||
AK145351.1 | - | 18 | 2875 | |||
AK145428.1 | - | 18 | 2692 | |||
AK145932.1 | - | 18 | 2686 | |||
AK146586.1 | - | 18 | 2710 | |||
AK146706.1 | - | 19 | 2794 | |||
AK147012.1 | - | 18 | 2708 | |||
AK150882.1 | - | 18 | 2687 | |||
AK151638.1 | - | 18 | 2705 | |||
AK151723.1 | - | 18 | 2709 | |||
AK151970.1 | - | 18 | 2709 | |||
AK152531.1 | - | 18 | 2705 | |||
AK152777.1 | - | 14 | 1916 | |||
AK153196.1 | - | 18 | 2686 | |||
AK153263.1 | - | 14 | 1916 | |||
AK154068.1 | - | 18 | 2694 | |||
AK155751.1 | - | 19 | 2787 | |||
AK159258.1 | - | 18 | 2702 | |||
AK159863.1 | - | 18 | 2702 | |||
AK168556.1 | - | 17 | 2506 | |||
BC016898.1 | - | 18 | 2706 | 4224466 | ||
BC024826.1 | - | 15 | 1989 | |||
BC029058.1 | - | 13 | 1648 | 4947555 | ||
BC029688.1 | - | 18 | 2705 | 5367783 | ||
BC031404.1 | - | 18 | 2677 | 4953772 | ||
BC036720.1 | - | 15 | 1995 | |||
BC055072.1 | - | 3705 | ||||
BC062097.1 | - | 19 | 2777 | 6833505 |