Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ahcyl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145542.3 | NP_663517.2 | 11, 12 | 1398 | ||
GenBank mRNA | AB092504.1 | - | 11, 12 | 1127 | ||
AK078823.1 | - | 4, 5 | 600 | |||
BC018218.1 | - | 11, 12 | 1227 | 4007102 |