Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADH1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000668.5 | NP_000659.2 | 2 | 212 | ||
NM_001286650.1 | NP_001273579.1 | 3 | 334 | |||
GenBank mRNA | AF153821.1 | - | 2 | 169 | ||
AK299409.1 | - | 2 | 169 | |||
AK301018.1 | - | 3 | 293 | |||
AK303945.1 | - | 2 | 171 | |||
AK304396.1 | - | 2 | 167 | |||
AK316501.1 | - | 2 | 169 | |||
BC033009.2 | - | 2 | 171 | 4828235 | ||
BP317995.1 | - | 2 | 185 | |||
BX641105.1 | - | 1 | 64 | |||
BX647469.1 | - | 2 | 2196 | |||
D00137.1 | - | 2 | 121 | |||
DC392123.1 | - | 2 | 214 | |||
FJ224285.1 | - | 2 | 171 | |||
M21692.1 | - | 2 | 133 | |||
M24317.1 | - | 2 | 131 | |||
X03350.1 | - | 2 | 173 |