Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc25a24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172685.3 | NP_766273.1 | 7 | 885 | ||
GenBank mRNA | AK032806.1 | - | 7 | 886 | ||
AK146034.1 | - | 7 | 884 | |||
AK167778.1 | - | 7 | 876 | |||
AK168090.1 | - | 7 | 876 | |||
AK170598.1 | - | 7 | 877 | |||
BC022637.1 | - | 2 | 100 | |||
BC055369.1 | - | 7 | 874 | 30055307 |