Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olfm3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286750.1 | NP_001273679.1 | 6 | 875 | ||
NM_153157.3 | NP_694797.1 | 6 | 1026 | |||
NM_153458.3 | NP_703188.1 | 6 | 1279 | |||
GenBank mRNA | AF442824.1 | - | 6 | 1026 | ||
AF442825.1 | - | 6 | 994 | |||
AK038569.1 | - | 6 | 1226 | |||
AK044325.1 | - | 6 | 1081 | |||
AK044605.1 | - | 6 | 1229 | |||
AK045482.1 | - | 6 | 1228 | |||
AK049612.1 | - | 6 | 1280 | |||
AK079315.1 | - | 6 | 1089 | |||
AK137414.1 | - | 6 | 1081 | |||
BC112422.1 | - | 6 | 935 | 40061966 | ||
BC113129.1 | - | 6 | 935 | 40061967 |