Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc35a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144902.3 | NP_659151.1 | 2 | 296 | ||
GenBank mRNA | AK029398.1 | - | 2 | 668 | ||
AK041195.1 | - | 3 | 416 | |||
AK049484.1 | - | 2 | 296 | |||
AK082040.1 | - | 2 | 274 | |||
AK161317.1 | - | 2 | 254 | |||
BC024110.1 | - | 2 | 292 | 5150304 |