Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccbl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001293560.1 | NP_001280489.1 | 3, 4 | 298 | ||
NM_173763.4 | NP_776124.1 | 3, 4 | 302 | |||
GenBank mRNA | AF363737.1 | - | 3, 4 | 272 | ||
AK049569.1 | - | 3, 4 | 271 | |||
AK145623.1 | - | 3, 4 | 296 | |||
BC085287.1 | - | 3, 4 | 253 | |||
BC131942.1 | - | 2, 3 | 87 | 40130219 | ||
BC132615.1 | - | 2, 3 | 87 | 40130892 |