Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam214b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253353.1 | NP_001240282.1 | 7, 8 | 1840 | ||
NM_001253354.1 | NP_001240283.1 | 8, 9 | 1617 | |||
NM_172691.3 | NP_766279.1 | 7, 8 | 1621 | |||
GenBank mRNA | AK045811.1 | - | 7, 8 | 1605 | ||
AK149610.1 | - | 7, 8 | 1698 | |||
AK154748.1 | - | 6, 7 | 1956 | |||
AK161380.1 | - | 7, 8 | 1548 | |||
AK173197.1 | - | 4, 5 | 366 | |||
BC022749.1 | - | 1, 2 | 33 | |||
BC044832.1 | - | 6, 7 | 1778 | |||
BC046771.1 | - | 8, 9 | 1607 | 5703628 | ||
BC054736.1 | - | 6, 7 | 1562 | |||
BC071191.1 | - | 5, 6 | 1309 | 6467750 |