Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Car9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139305.2 | NP_647466.2 | 7 | 951 | ||
GenBank mRNA | AJ245857.1 | - | 7 | 953 | ||
AK136579.1 | - | 7 | 952 | |||
AK137719.1 | - | 7 | 953 | |||
AK166403.1 | - | 4 | 380 | |||
BC120544.1 | - | 7 | 944 | 40129467 | ||
BC120546.1 | - | 7 | 944 | 40129469 |