Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Galnt12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172693.3 | NP_766281.1 | 9 | 1491 | ||
GenBank mRNA | AB175680.1 | - | 9 | 1471 | ||
AK033638.1 | - | 9 | 1492 | |||
AK042133.1 | - | 9 | 1524 | |||
AK043977.1 | - | 8 | 1352 | |||
AK082014.1 | - | 9 | 1537 | |||
AK156751.1 | - | 9 | 1521 | |||
BC024475.1 | - | 8 | 1323 | |||
BC025639.1 | - | 8 | 1213 | |||
BC056425.1 | - | 9 | 1502 | 5716909 |