Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmeff1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021436.2 | NP_067411.1 | 9 | 1200 | ||
GenBank mRNA | AJ400622.1 | - | 9 | 869 | ||
AK043792.1 | - | 7 | 576 | |||
AK079633.1 | - | 7 | 550 | |||
BC023714.1 | - | 4 | 362 | |||
BC057598.1 | - | 9 | 1200 | 6401547 |