Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acnat1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164565.1 | NP_001158037.1 | 3 | 978 | ||
GenBank mRNA | DQ469311.1 | - | 3 | 978 |