Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aldob | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144903.2 | NP_659152.1 | 8 | 910 | ||
GenBank mRNA | AK135385.1 | - | 8 | 939 | ||
AK149395.1 | - | 8 | 927 | |||
AK149598.1 | - | 8 | 924 | |||
AK167334.1 | - | 8 | 927 | |||
AK167566.1 | - | 8 | 924 | |||
AK168600.1 | - | 8 | 924 | |||
AK168997.1 | - | 8 | 974 | |||
BC016435.1 | - | 8 | 906 | 4241025 | ||
BC022113.1 | - | 8 | 883 | 5098524 | ||
BC024056.1 | - | 8 | 916 | 5100067 | ||
BC024112.1 | - | 8 | 915 | 5102118 | ||
BC026577.1 | - | 8 | 875 | 4989521 | ||
BC030724.1 | - | 8 | 912 | 5100784 | ||
BC030725.1 | - | 8 | 916 | 5101686 | ||
BC034169.1 | - | 8 | 916 | 4975382 | ||
BC034171.1 | - | 8 | 893 | 4972521 | ||
BC034172.1 | - | 8 | 880 | 4974489 | ||
BC034173.1 | - | 8 | 892 | 4974553 | ||
BC036130.1 | - | 8 | 893 | 4974071 | ||
BC036131.1 | - | 8 | 910 | 4974877 | ||
BC036132.1 | - | 8 | 910 | 4975336 | ||
BC036133.1 | - | 8 | 922 | 4975573 | ||
BC036139.1 | - | 4 | 534 | |||
BC036142.1 | - | 2 | 217 |