Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Haus6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173400.2 | NP_775576.2 | 11 | 1425 | ||
GenBank mRNA | AK036635.1 | - | 2 | 90 | ||
AK051016.1 | - | 11 | 1446 | |||
AK077681.1 | - | 11 | 1447 | |||
AK136163.1 | - | 2 | 145 | |||
AK169222.1 | - | 11 | 1427 | |||
AK173206.1 | - | 11 | 1397 | |||
BC064725.1 | - | 6 | 622 | |||
BC068237.1 | - | 11 | 1425 | 30550558 |