Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Haus6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173400.2 | NP_775576.2 | 16 | 2307 | ||
GenBank mRNA | AK036635.1 | - | 7 | 972 | ||
AK077681.1 | - | 16 | 2329 | |||
AK136163.1 | - | 7 | 1028 | |||
AK169222.1 | - | 16 | 2309 | |||
AK173206.1 | - | 15 | 3733 | |||
BC044803.1 | - | 1 | 68 | |||
BC064725.1 | - | 11 | 1504 | |||
BC068237.1 | - | 16 | 2307 | 30550558 |