Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acer2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290541.1 | NP_001277470.1 | 3 | 419 | ||
NM_001290543.1 | NP_001277472.1 | 3 | 419 | |||
NM_139306.3 | NP_647467.1 | 3 | 419 | |||
GenBank mRNA | AF282864.1 | - | 3 | 378 | ||
AK085306.1 | - | 3 | 415 | |||
AY312515.1 | - | 3 | 344 | |||
BC059819.1 | - | 3 | 419 | 6839525 |