Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pars2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083887.2 | NP_001077356.1 | 2 | 1862 | ||
NM_001285783.1 | NP_001272712.1 | 2 | 1879 | |||
NM_001285784.1 | NP_001272713.1 | 2 | 1030 | |||
NM_172272.2 | NP_758476.2 | 2 | 1013 | |||
GenBank mRNA | BC027073.1 | - | 1 | 703 |