Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADH1C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000669.4 | NP_000660.1 | 6 | 1168 | ||
XM_011531588.2 | XP_011529890.1 | 5 | 958 | |||
XM_011531589.1 | XP_011529891.1 | 7 | 1288 | |||
GenBank mRNA | BC062476.1 | - | 6 | 864 | 30408761 | |
BC066227.1 | - | 6 | 826 | 7002195 | ||
BC066228.1 | - | 6 | 826 | 7002197 | ||
BC067419.1 | - | 6 | 826 | 7002191 | ||
BC067420.1 | - | 6 | 826 | 7002192 | ||
BC067421.1 | - | 6 | 826 | 7002193 | ||
BC067422.1 | - | 6 | 826 | 7002194 | ||
BC074771.2 | - | 6 | 826 | 30915457 | ||
BC074786.2 | - | 6 | 825 | 30915344 | ||
M12272.1 | - | 6 | 878 | |||
X04299.1 | - | 6 | 870 | |||
X04350.1 | - | 4 | 640 |
Gene Symbol: ADH1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000668.5 | NP_000659.2 | 6 | 909 | ||
NM_001286650.1 | NP_001273579.1 | 7 | 1031 | |||
GenBank mRNA | AF153821.1 | - | 6 | 866 | ||
AK299409.1 | - | 7 | 827 | |||
AK301018.1 | - | 7 | 990 | |||
AK303945.1 | - | 5 | 699 | |||
AK304396.1 | - | 6 | 633 | |||
AK316501.1 | - | 6 | 699 | |||
BC033009.2 | - | 6 | 868 | 4828235 | ||
BX641105.1 | - | 1 | 5017 | |||
BX647469.1 | - | 6 | 2893 | |||
D00137.1 | - | 6 | 818 | |||
FJ224285.1 | - | 6 | 868 | |||
M21692.1 | - | 6 | 830 | |||
M24317.1 | - | 6 | 828 | |||
X03350.1 | - | 6 | 870 |