Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrrc41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK028000.1 | - | 5 | 616 | ||
AK052762.1 | - | 8 | 1919 | |||
AK154613.1 | - | 8 | 2065 | |||
BC005765.1 | - | 7 | 1786 | |||
BC007169.1 | - | 5 | 944 | |||
BC022976.1 | - | 8 | 1997 | 5362925 | ||
BC024940.1 | - | 8 | 1994 | |||
BC071258.1 | - | 8 | 1930 |