Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLGLB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002665.4 | NP_002656.1 | 4 | 1075 | ||
GenBank mRNA | AK124365.1 | - | 3 | 1979 | ||
BC005379.1 | - | 4 | 1071 | 3934860 | ||
M93143.2 | - | 4 | 1075 |
Gene Symbol: PLGLB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001032392.2 | NP_001027564.1 | 4 | 1071 | ||
GenBank mRNA | BC070296.1 | - | 4 | 1050 | ||
BC107767.1 | - | 4 | 1046 |