Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLGLB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001032392.2 | NP_001027564.1 | 4 | 1258 |
Gene Symbol: PLGLB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002665.4 | NP_002656.1 | 4 | 1262 | ||
GenBank mRNA | AF075095.1 | - | 1 | 144 |