Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Epha10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256432.1 | NP_001243361.1 | 3 | 481 | ||
NM_177671.5 | NP_808339.2 | 3 | 481 | |||
GenBank mRNA | AK039700.1 | - | 3 | 444 | ||
AK143168.1 | - | 1366 | ||||
BC116991.1 | - | 3 | 411 | 40126310 | ||
BC119084.1 | - | 3 | 411 | 8733833 |