Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADH1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000667.3 | NP_000658.1 | 2 | 170 | ||
GenBank mRNA | AK290558.1 | - | 2 | 127 | ||
AK309597.1 | - | 2 | 145 | |||
BC074738.2 | - | 2 | 84 | 30915195 | ||
BC117442.1 | - | 2 | 126 | 40125993 | ||
BC126306.1 | - | 2 | 126 | 8992022 | ||
BT019812.1 | - | 2 | 56 | |||
CB116632.1 | - | 2 | 170 | |||
M12271.1 | - | 2 | 71 | |||
M12963.1 | - | 2 | 128 |
Gene Symbol: ADH1C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000669.4 | NP_000660.1 | 2 | 426 | ||
NR_133005.1 | 2 | 426 | ||||
XM_011531589.1 | XP_011529891.1 | 3 | 546 | |||
GenBank mRNA | BC062476.1 | - | 2 | 122 | 30408761 | |
BC066227.1 | - | 2 | 84 | 7002195 | ||
BC066228.1 | - | 2 | 84 | 7002197 | ||
BC067419.1 | - | 2 | 84 | 7002191 | ||
BC067420.1 | - | 2 | 84 | 7002192 | ||
BC067421.1 | - | 2 | 84 | 7002193 | ||
BC067422.1 | - | 2 | 84 | 7002194 | ||
BC074771.2 | - | 2 | 84 | 30915457 | ||
BC074786.2 | - | 2 | 83 | 30915344 | ||
CB114902.1 | - | 2 | 141 | |||
CD014078.1 | - | 2 | 127 | |||
DA562068.1 | - | 3 | 546 | |||
M12272.1 | - | 2 | 136 | |||
X04299.1 | - | 2 | 128 |