Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zbtb40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198248.1 | NP_937891.1 | 6 | 1274 | ||
GenBank mRNA | AK220256.1 | - | 7 | 1159 | ||
BC059177.1 | - | 6 | 1274 | 6410689 |