Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FXYD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136007.1 | NP_001129479.1 | 7 | 534 | ||
NM_001136008.1 | NP_001129480.1 | 5 | 328 | |||
NM_001136009.1 | NP_001129481.1 | 6 | 412 | |||
NM_001136010.1 | NP_001129482.1 | 5 | 328 | |||
NM_001136011.1 | NP_001129483.1 | 6 | 412 | |||
NM_001136012.1 | NP_001129484.1 | 5 | 328 | |||
NM_005971.3 | NP_005962.1 | 5 | 328 | |||
NM_021910.2 | NP_068710.1 | 6 | 412 | |||
XM_005258996.4 | XP_005259053.1 | 4 | 233 | |||
XM_011527032.2 | XP_011525334.1 | 7 | 541 | |||
XM_017026877.1 | XP_016882366.1 | 5 | 278 | |||
XM_017026878.1 | XP_016882367.1 | 4 | 198 | |||
GenBank mRNA | BC005238.1 | - | 4 | 142 | 3997821 | |
BC090044.1 | - | 5 | 277 | 30395606 | ||
BE793135.1 | - | 3 | 102 | |||
BI715233.1 | - | 4 | 118 | |||
BM549935.1 | - | 3 | 140 | |||
BQ676323.1 | - | 6 | 360 | |||
BQ682674.1 | - | 6 | 658 | |||
BT006712.1 | - | 3 | 86 | |||
BU185141.1 | - | 5 | 215 | |||
CA948888.1 | - | 2 | 55 | |||
CB128589.1 | - | 4 | 141 | |||
CR456945.1 | - | 3 | 86 | |||
CR542197.1 | - | 3 | 86 | |||
DA438143.1 | - | 4 | 142 | |||
DA890604.1 | - | 4 | 170 | |||
DB202948.1 | - | 4 | 143 | |||
U28249.1 | - | 6 | 345 | |||
X93036.1 | - | 4 | 145 |