Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxo44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161851.2 | NP_001155323.1 | 2 | 148 | ||
NM_001161852.2 | NP_001155324.1 | 2 | 425 | |||
NM_173401.5 | NP_775577.2 | 2 | 425 | |||
GenBank mRNA | AK019407.1 | - | 2 | 181 | ||
AK077452.1 | - | 2 | 211 | |||
AK170958.1 | - | 2 | 108 | |||
AK177996.1 | - | 2 | 141 | |||
AK178346.1 | - | 2 | 140 | |||
AK191511.1 | - | 1 | 21 | |||
BC028884.1 | - | 2 | 217 | 4165174 | ||
BM932876.1 | - | 1 | 74 | |||
BQ042735.1 | - | 2 | 208 | |||
BY252242.1 | - | 2 | 135 | |||
BY292524.1 | - | 2 | 209 |