Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxo44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161851.2 | NP_001155323.1 | 2 | 224 | ||
NM_001161852.2 | NP_001155324.1 | 2 | 501 | |||
NM_173401.5 | NP_775577.2 | 2 | 501 | |||
GenBank mRNA | AK019407.1 | - | 2 | 257 | ||
AK077452.1 | - | 2 | 287 | |||
AK170958.1 | - | 2 | 184 | |||
AK177996.1 | - | 2 | 217 | |||
AK191511.1 | - | 1 | 97 | |||
AK218805.1 | - | 1 | 30 | |||
BC028884.1 | - | 2 | 293 | 4165174 | ||
BM932876.1 | - | 1 | 150 | |||
BQ042735.1 | - | 2 | 284 | |||
BY252242.1 | - | 2 | 211 |