Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gckr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144909.1 | NP_659158.1 | 14, 15 | 1273 | ||
GenBank mRNA | AK149377.1 | - | 12, 13 | 3172 | ||
BC012412.1 | - | 14, 15 | 1273 | 4159892 |