Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc10a4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173403.2 | NP_775579.2 | 3 | 1842 | ||
GenBank mRNA | AK087479.1 | - | 3 | 1843 | ||
AK149202.1 | - | 1843 | ||||
BC132529.1 | - | 3 | 1754 | 40130806 | ||
BC138048.1 | - | 3 | 1754 | 8861067 |