Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Polr2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153798.2 | NP_722493.2 | 2 | 206 | ||
GenBank mRNA | AK039766.1 | - | 2 | 207 | ||
BC016415.1 | - | 2 | 194 | |||
BC038472.1 | - | 2 | 199 | 5026936 |