Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ythdc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177680.3 | NP_808348.2 | 6 | 1093 | ||
GenBank mRNA | AK173312.1 | - | 3 | 356 | ||
CN526522.1 | - | 3 | 417 | |||
CO424360.1 | - | 1 | 38 |