Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Grsf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098476.2 | NP_001091946.1 | 3 | 255 | ||
NM_178700.5 | NP_848815.2 | 3 | 583 | |||
GenBank mRNA | AK044889.1 | - | 3 | 584 | ||
AK045986.1 | - | 3 | 537 | |||
AK077782.1 | - | 3 | 584 | |||
AK089014.1 | - | 2 | 119 | |||
AK168385.1 | - | 3 | 226 | |||
AK168459.1 | - | 3 | 226 | |||
CK391158.1 | - | 3 | 243 |