Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Parm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145562.2 | NP_663537.1 | 2 | 931 | ||
GenBank mRNA | AK078862.1 | - | 2 | 893 | ||
AK078943.1 | - | 2 | 945 | |||
AK148300.1 | - | 2 | 979 | |||
AK161172.1 | - | 894 | ||||
AK167402.1 | - | 2 | 920 | |||
BC006604.1 | - | 2 | 931 | 3966312 | ||
EF564364.1 | - | 2 | 716 |