Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Parm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145562.2 | NP_663537.1 | 3 | 1004 | ||
GenBank mRNA | AK078862.1 | - | 3 | 966 | ||
AK078943.1 | - | 3 | 1018 | |||
AK148300.1 | - | 3 | 1052 | |||
AK161172.1 | - | 967 | ||||
AK167402.1 | - | 3 | 993 | |||
BC006604.1 | - | 3 | 1004 | 3966312 | ||
EF564364.1 | - | 3 | 789 |