Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sdad1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172713.2 | NP_766301.1 | 4 | 481 | ||
GenBank mRNA | AK029881.1 | - | 4 | 483 | ||
AK160646.1 | - | 4 | 352 | |||
AK164608.1 | - | 4 | 447 | |||
BC042708.1 | - | 4 | 431 | 5024980 |