Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Paqr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198422.2 | NP_940814.1 | 2 | 356 | ||
GenBank mRNA | AK149213.1 | - | 2 | 335 | ||
AK158711.1 | - | 2 | 344 | |||
AK172689.1 | - | 2 | 311 | |||
AY424292.1 | - | 2 | 356 | |||
BC050248.1 | - | 2 | 356 | |||
BC051173.1 | - | 2 | 356 | |||
BC120853.1 | - | 2 | 341 | 40129776 | ||
BC120855.1 | - | 2 | 342 | 40129778 |