Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: A830010M20Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007574.2 | NP_001007575.2 | 4 | 907 | ||
NM_001168557.1 | NP_001162029.1 | 4 | 873 | |||
GenBank mRNA | AK043587.1 | - | 4 | 908 | ||
AK043720.1 | - | 4 | 875 | |||
AK122444.1 | - | 4 | 856 | |||
AK164388.1 | - | 4 | 877 | |||
AK169666.1 | - | 4 | 874 |