Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ficd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010825.3 | NP_001010825.2 | 3 | 767 | ||
GenBank mRNA | AK044446.1 | - | 3 | 576 | ||
AK054325.1 | - | 3 | 532 | |||
AK087477.1 | - | 3 | 751 | |||
BC106090.1 | - | 3 | 738 | |||
BC139821.1 | - | 2 | 404 | 40141450 |